Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PSMB1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556219
Description
Immunogen sequence: SGFHGDCLTL TKIIEARLKM YKHSNNKAMT TGAIAAMLST ILYSRRFFPY YVYNIIGGLD EEGKGAVYSF DPVGS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
PSMB1 is a multicatalytic proteinase complex which is characterized by its ability to cleave peptides with Arg, Phe, Tyr, Leu, and Glu adjacent to the leaving group at neutral or slightly basic pH. The proteasome has an ATP-dependent proteolytic activity.
Specifications
| PSMB1 | |
| Polyclonal | |
| Unconjugated | |
| PSMB1 | |
| AA409053; C81484; component C5 of proteasome; HC5; Lmpc5; Macropain subunit C5; multicatalytic endopeptidase complex subunit C5; PMSB1; proteasome (prosome, macropain) subunit, beta type 1; proteasome (prosome, macropain) subunit, beta type, 1; proteasome beta 1 subunit; proteasome beta-subunit C5; proteasome component C5; Proteasome gamma chain; proteasome subunit beta 1; proteasome subunit beta type-1; proteasome subunit C5; proteasome subunit HC5; proteasome subunit RC5; PSC5; Psmb1; testicular secretory protein Li 45 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5689 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P20618 | |
| PSMB1 | |
| Recombinant protein corresponding to Human PSMB1. Recombinant protein control fragment (Product #RP-94588). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction