Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSMB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578135
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578135 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578135 Supplier Invitrogen™ Supplier No. PA578135
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: 293T, A431 Predicted Reactivity: Zebrafish (80%), Pig (97%), Chicken (85%), Rhesus Monkey (98%), Bovine (95%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

PSMB1 is a multicatalytic proteinase complex which is characterized by its ability to cleave peptides with Arg, Phe, Tyr, Leu, and Glu adjacent to the leaving group at neutral or slightly basic pH. The proteasome has an ATP-dependent proteolytic activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMB1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.89 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene PSMB1
Gene Accession No. O09061, P18421, P20618
Gene Alias AA409053; C81484; component C5 of proteasome; HC5; Lmpc5; Macropain subunit C5; multicatalytic endopeptidase complex subunit C5; PMSB1; proteasome (prosome, macropain) subunit, beta type 1; proteasome (prosome, macropain) subunit, beta type, 1; proteasome beta 1 subunit; proteasome beta-subunit C5; proteasome component C5; Proteasome gamma chain; proteasome subunit beta 1; proteasome subunit beta type-1; proteasome subunit C5; proteasome subunit HC5; proteasome subunit RC5; PSC5; Psmb1; testicular secretory protein Li 45
Gene Symbols PSMB1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PSMB1. The exact sequence is proprietary.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19170, 5689, 94198
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.