Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSMB10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596327
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596327 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596327 Supplier Invitrogen™ Supplier No. PA596327
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: THP-1; Cellular Location: Cytoplasm, Nucleus.

The 20S proteasome is a protease complex that is responsible for cytosolic protein degradation and generation of peptide ligands for major histocompatibility complex (MHC) class I molecules, either in their final form or in the form of amino-terminally extended precursors. Upon IFN-gamma stimulation of cells, three constitutively expressed subunits of the 20S proteasome are replaced by inducible subunits LMP2 (low-molecular mass polypeptide 2), LMP7, and MECL-1 (multicatalytic endopeptidase complex-like-1, LMP10). LMP2, LMP7, and MECL-1 subunits form immunoproteasomes, which are associated with more efficient class I antigen processing and presentation. Independent assortment of LMP-2, LMP-7, and MECL-1 into different proteasome complexes can lead to 36 unique proteasome subsets, which may mediate differences in the cleavage specificities/cleavage motifs of proteins subject to constitutive- and immuno-proteasomes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMB10
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.97 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PSMB10
Gene Accession No. O35955, P40306, Q4KM35
Gene Alias beta2i; LMP10; Low molecular mass protein 10; macropain; Macropain subunit MECl-1; MECL1; Mecl-1; MGC1665; multicatalytic endopeptidase complex subunit MECl-1; OTTHUMP00000174858; prosome; prosome Mecl1; proteasome (prosome, macropain) subunit, beta type 10; proteasome (prosome, macropain) subunit, beta type, 10; proteasome (prosomome, macropain) subunit, beta type 10; proteasome 20S subunit beta 10; proteasome beta 10 subunit; proteasome catalytic subunit 2i; Proteasome MECl 1; proteasome MECl-1; proteasome subunit 10; proteasome subunit beta 10; proteasome subunit beta 7i; proteasome subunit beta type-10; Proteasome subunit beta-2i; proteasome subunit LMP10; proteasome subunit MECL1; PSMB 10; Psmb10
Gene Symbols PSMB10
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19171, 291983, 5699
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.