Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSMB11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563376
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563376 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563376 Supplier Invitrogen™ Supplier No. PA563376
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GSVDLFHVRE SGWEHVSRSD ACVLYVELQK LLEPEPEEDA SHAHPEPATA HRAAEDRELS VGPGEVTPGD SRMPAGTET Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 62%.

Proteasomes generate peptides that are presented by major histocompatibility complex. Three additional subunits, beta-1i, beta-2i, and beta-5i, are induced by gamma-interferon and are preferentially incorporated into proteasomes to make immunoproteasomes. PSMB11, or beta-5t, is a catalytic subunit expressed exclusively in cortical thymic epithelial cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMB11
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PSMB11
Gene Accession No. A5LHX3
Gene Alias 5830406J20Rik; beta5t; proteasome (prosome, macropain) subunit, beta type, 11; proteasome beta 11 subunit; proteasome subunit beta 11; proteasome subunit beta 5T; proteasome subunit beta type-11; proteasome subunit beta5t; Proteasome subunit beta-5t; Psmb11; RGD1308773
Gene Symbols PSMB11
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PSMB11. Recombinant protein control fragment (Product #RP-101630).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 122706
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.