Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PSMB2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597970
Description
PSMB2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
The proteasome is a multicatalytic proteinase complex which is characterized by its ability to cleave peptides with Arg, Phe, Tyr, Leu, and Glu adjacent to the leaving group at neutral or slightly basic pH. The proteasome has an ATP-dependent proteolytic activity. This subunit has a trypsin-like activity.
Specifications
PSMB2 | |
Polyclonal | |
Unconjugated | |
PSMB2 | |
AU045357; AW108089; C7-I; D4Wsu33e; HC7-I; Macropain subunit C7-I; multicatalytic endopeptidase complex subunit C7-1; multicatalytic endopeptidase complex subunit C7-I; proteasome (prosome, macropain) subunit, beta type 2; proteasome (prosome, macropain) subunit, beta type, 2; proteasome beta 2 subunit; proteasome component C7-I; proteasome subunit beta 2; proteasome subunit beta type-2; proteasome subunit, beta type, 2; PSMB2; testicular tissue protein Li 152 | |
Rabbit | |
Protein G | |
RUO | |
26445, 5690 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
P49721, Q9R1P3 | |
PSMB2 | |
Recombinant Human Proteasome subunit beta type-2 protein (1-201AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction