Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSMB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555304
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555304 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555304 Supplier Invitrogen™ Supplier No. PA555304
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLLAGYDEHE GPALYYMDYL AALAKAPFAA HGYGAFLTLS ILDRYYTPTI SRERAVELLR KCLEELQKRF ILNLPTF Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

The proteasome is a multicatalytic proteinase complex which is characterized by its ability to cleave peptides with Arg, Phe, Tyr, Leu, and Glu adjacent to the leaving group at neutral or slightly basic pH. The proteasome has an ATP-dependent proteolytic activity. This subunit has a trypsin-like activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMB2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PSMB2
Gene Accession No. P40307, P49721, Q9R1P3
Gene Alias AU045357; AW108089; C7-I; D4Wsu33e; HC7-I; Macropain subunit C7-I; multicatalytic endopeptidase complex subunit C7-1; multicatalytic endopeptidase complex subunit C7-I; proteasome (prosome, macropain) subunit, beta type 2; proteasome (prosome, macropain) subunit, beta type, 2; proteasome beta 2 subunit; proteasome component C7-I; proteasome subunit beta 2; proteasome subunit beta type-2; proteasome subunit, beta type, 2; PSMB2; testicular tissue protein Li 152
Gene Symbols PSMB2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PSMB2. Recombinant protein control fragment (Product #RP-94170).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26445, 29675, 5690
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.