Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSMB2 Recombinant Rabbit Monoclonal Antibody (PSH0-28)
GREENER_CHOICE

Catalog No. FEMA550799
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550799 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550799 Supplier Invitrogen™ Supplier No. MA550799
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive control: Jurkat cell lysate, HepG2 cell lysate, HeLa cell lysate, HEK-293 cell lysate, SK-OV-3 cell lysate, HT-29 cell lysate, PC-3M cell lysate. Predicted band size: 23 kDa Subcellular Location: Cytoplasm, Nucleus.

The proteasome is a multicatalytic proteinase complex which is characterized by its ability to cleave peptides with Arg, Phe, Tyr, Leu, and Glu adjacent to the leaving group at neutral or slightly basic pH. The proteasome has an ATP-dependent proteolytic activity. This subunit has a trypsin-like activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMB2
Applications Western Blot
Classification Recombinant Monoclonal
Clone PSH0-28
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene PSMB2
Gene Accession No. P49721
Gene Alias AU045357; AW108089; C7-I; D4Wsu33e; HC7-I; Macropain subunit C7-I; multicatalytic endopeptidase complex subunit C7-1; multicatalytic endopeptidase complex subunit C7-I; proteasome (prosome, macropain) subunit, beta type 2; proteasome (prosome, macropain) subunit, beta type, 2; proteasome beta 2 subunit; proteasome component C7-I; proteasome subunit beta 2; proteasome subunit beta type-2; proteasome subunit, beta type, 2; PSMB2; testicular tissue protein Li 152
Gene Symbols PSMB2
Host Species Rabbit
Immunogen Synthetic peptide within Human PSMB2 aa 175-201/201.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5690
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.