Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSMB4 Mouse anti-Human, Clone: OTI2C9, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA503558

Description
The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the proteasome B-type family, also known as the T1B family, that is a 20S core beta subunit.Specifications
PSMB4 | |
Monoclonal | |
0.59 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P28070 | |
PSMB4 | |
Full length human recombit protein of human PSMB4 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2C9 | |
Unconjugated | |
PSMB4 | |
HN3, HsN3, PROS-26, PROS26 | |
Mouse | |
Affinity Chromatography | |
RUO | |
5692 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction