Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMB4 Monoclonal Antibody (OTI6F10), TrueMAB™, OriGene
SDP

Mouse Monoclonal Antibody

Supplier:  OriGene TA503617

Encompass_Preferred

 View more versions of this product

Catalog No. 50-167-4470


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the proteasome B-type family, also known as the T1B family, that is a 20S core beta subunit.
Specifications

Specifications

PSMB4
Monoclonal
0.23 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P28070
PSMB4
Full length human recombit protein of human PSMB4 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG2b
Western Blot
OTI6F10
Unconjugated
PSMB4
HN3, HsN3, PROS-26, PROS26
Mouse
Affinity Chromatography
RUO
5692
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.