Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMB4 Monoclonal Antibody (OTI6F10), TrueMAB™, OriGene
SDP

Mouse Monoclonal Antibody

Supplier:  OriGene TA503617S

Encompass
Encompass_Preferred

 View more versions of this product

Catalog No. 50-254-0798


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

PSMB4 Monoclonal antibody specifically detects PSMB4 in Human, Mouse samples. It is validated for Western Blot
Specifications

Specifications

PSMB4
Monoclonal
0.23 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9EQ80, Q9GZT8
Mouse
Affinity chromatography
RUO
60491, 65102
-20° C, Avoid Freeze/Thaw Cycles
IgG2b
Western Blot
OTI6F10
Unconjugated
NIF3L1
26 kDa prosomal protein; HN3; HsBPROS26; HsN3; Lmp3; Low molecular mass protein 3; macropain beta chain; Multicatalytic endopeptidase complex beta chain; PROS26; PROS-26; Pros-27; proteasome (prosome, macropain) subunit, beta type 4; proteasome (prosome, macropain) subunit, beta type, 4; proteasome beta 4 subunit; proteasome beta chain; Proteasome chain 3; proteasome subunit beta 4; proteasome subunit beta type-4; proteasome subunit HsN3; proteasome subunit, beta type, 4; Psmb4; RN3; testis tissue sperm-binding protein Li 79P
Full length human recombinant protein of human NIF3L1 produced in HEK293T cell.
30 μL
Primary
Human, Mouse
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.