Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMB4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102828
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102828 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102828 Supplier Thermo Scientific Supplier No. PA5102828
Add to Cart
Add to Cart

PSMB4 Polyclonal Antibody

PSMB4 Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMB4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PSMB4
Gene Accession No. P28070, P34067, P99026
Gene Alias 26 kDa prosomal protein; HN3; HsBPROS26; HsN3; Lmp3; Low molecular mass protein 3; macropain beta chain; Multicatalytic endopeptidase complex beta chain; PROS26; PROS-26; Pros-27; proteasome (prosome, macropain) subunit, beta type 4; proteasome (prosome, macropain) subunit, beta type, 4; proteasome beta 4 subunit; proteasome beta chain; Proteasome chain 3; proteasome subunit beta 4; proteasome subunit beta type-4; proteasome subunit HsN3; proteasome subunit, beta type, 4; Psmb4; RN3; testis tissue sperm-binding protein Li 79P
Gene Symbols PSMB4
Host Species Rabbit
Immunogen A synthesized peptide derived from human PSMB4(Accession P28070), corresponding to amino acid residues E120-V170.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19172, 5692, 58854
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.