Learn More
Invitrogen™ PSMB5 Polyclonal Antibody

Description
Recommended positive controls: A431, H1299. Predicted reactivity: Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | PSMB5 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | PSMB5 |
| Gene Accession No. | P28074 |
| Gene Alias | LMPX; Macropain epsilon chain; MB1; MGC118075; Multicatalytic endopeptidase complex epsilon chain; proteasome (prosome, macropain) subunit, beta type 5; proteasome (prosome, macropain) subunit, beta type, 5; proteasome 20S subunit beta 5; proteasome beta 5 subunit; proteasome beta type subunit 5; proteasome catalytic subunit 3; proteasome chain 6; Proteasome epsilon chain; proteasome subunit beta 5; proteasome subunit beta type-5; Proteasome subunit MB1; proteasome subunit X; proteasome subunit, beta type, 5; PSMB5; PSX large multifunctional protease X; testicular tissue protein Li 153; Unknown (protein for MGC:134464); X |
| Gene Symbols | PSMB5 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.