Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSMB6 Polyclonal Antibody

Catalog No. PIPA5121890
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121890 20 μL
PIPA5119367 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121890 Supplier Invitrogen™ Supplier No. PA5121890
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PSMB6 Polyclonal Antibody for Western Blot, IHC (P)

Proteolytic degradation is critical to the maintenance of appropriate levels of short-lived and regulatory proteins as important and diverse as those involved in cellular metabolism, heat shock and stress response, antigen presentation, modulation of cell surface receptors and ion channels, cell cycle regulation, transcription, and signaling factors. The ubiquitin-proteasome pathway deconstructs most proteins in the eukaryotic cell cytosol and nucleus. Other proteins are degraded via the vacuolar pathway which includes endosomes, lysosomes, and the endoplasmic reticulum. The 26S proteasome is an ATP-dependent, multisubunit (~31), barrel-shaped molecular machine with an apparent molecular weight of ~2.5 MDa. It consists of a 20S proteolytic core complex which is crowned at one or both ends by 19S regulatory subunit complexes. The 19S regulatory subunits recognize ubiquitinated proteins and play an essential role in unfolding and translocating targets into the lumen of the 20S subunit. The PA28/11S REG Activator protein complex functions as a proteolytic activator. This complex, consisting of alpha, beta, and gamma subunits, enhances the activity of the 20S proteolytic core. An enzymatic cascade is responsible for the attachment of multiple ubiquitin molecules to lysine residues of proteins targeted for degradation. Several genetic diseases are associated with defects in the ubiquitin-proteasome pathway. Some examples of affected proteins include those linked to cystic fibrosis (CF transmembrane regulator), Angelmane's syndrome (E6-AP), and Liddle syndrome (endothelial sodium channels).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMB6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene PSMB6
Gene Accession No. P28072
Gene Alias DELTA; Lmp19; LMPY; low molecular mass protein 19; Macropain delta chain; Mpnd; multicatalytic endopeptidase complex delta chain; proteasome (prosome, macropain) subunit, beta type 6; proteasome (prosome, macropain) subunit, beta type, 6; proteasome 20S subunit beta 6; proteasome beta 6 subunit; proteasome catalytic subunit 1; proteasome chain 5; Proteasome delta chain; proteasome subunit beta 6; proteasome subunit beta type-6; proteasome subunit delta; proteasome subunit Y; Psmb6; Psmb6l; PSY large multifunctional protease Y; RP23-122P1.4; subunit 2; Y
Gene Symbols PSMB6
Host Species Rabbit
Immunogen Recombinant protein Proteasome Subunit Beta Type 6. The antigen corresponds to amino acid range 2-239 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5694
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.