Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSMB7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP25864825UL
Description
PSMB7 Polyclonal specifically detects PSMB7 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Proteasome 20S beta 7 | |
Polyclonal | |
Western Blot 0.4 μg/mL, Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
EC 3.4.25.1, Macropain chain Z, Multicatalytic endopeptidase complex chain Z, proteasome (prosome, macropain) subunit, beta type, 7, proteasome subunit alpha, proteasome subunit beta 7, proteasome subunit beta type-7, Proteasome subunit Z, PSMB7, Zproteasome catalytic subunit 2 | |
Rabbit | |
Affinity Purified | |
RUO | |
5695 | |
Human | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
PSMB7 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LVSEAIAAGIFNDLGSGSNIDLCVISKNKLDFLRPYTVPNKKGTRLGRYRCEKGTTAVLTEKITPLEIEVLEETVQTM | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction