Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSMB7 Antibody (OTI1E1), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273645FR
Description
PSMB7 Monoclonal specifically detects PSMB7 in Human, Mouse, Rat, Monkey, Primate samples. It is validated for Western Blot, Immunohistochemistry.Specifications
PSMB7 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat, Monkey, Primate | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1E1 | |
Western Blot, Immunohistochemistry | |
PSMB7 | |
Recombinant protein fragment corresponding to amino acids 58-277 of human Proteasome 20S beta 7 (NP_002790) produced in E.coli. | |
0.1 mL | |
5695.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction