Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSMB7 Antibody (OTI1E1), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273645PCP
Description
PSMB7 Monoclonal antibody specifically detects PSMB7 in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
PSMB7 | |
Monoclonal | |
PerCP | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Primate, Monkey | |
Purified |
Western Blot, Immunohistochemistry | |
OTI1E1 | |
Western Blot, Immunohistochemistry | |
Z | |
Human recombinant protein fragment corresponding to amino acids 58-277 of human PSMB7(NP_002790) produced in E.coli. | |
0.1 mL | |
Neuroscience, Ubiquitin Proteasome Pathway | |
5695 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction