Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMB7 Monoclonal Antibody (OTI4E3), TrueMAB™, OriGene
SDP

Catalog No. 502540201
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0201 30 μL
50-167-5285 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0201 Supplier OriGene Supplier No. TA504638S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PSMB7 Monoclonal antibody specifically detects PSMB7 in Human samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen PSMB7
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI4E3
Concentration 0.83 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene TDO2
Gene Accession No. P48775
Gene Alias AU020723; beta-type proteasome 7 subunit; EGK_07890; hypothetical protein LOC511207; macropain chain Z; MC14; Mmc14; Multicatalytic endopeptidase complex chain Z; proteasome (prosome, macropain) subunit, beta type 7; proteasome (prosome, macropain) subunit, beta type, 7; proteasome 20S subunit beta 7; proteasome beta 7 subunit; proteasome catalytic subunit 2; proteasome subunit alpha; proteasome subunit beta 7; proteasome subunit beta type-7; proteasome subunit beta type-7 proprotein; proteasome subunit Z; protein serine kinase c17; Psmb7; RP11-101K10.7; Z
Host Species Mouse
Immunogen Full length human recombinant protein of human TDO2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6999
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.