Learn More
Invitrogen™ PSMB7 Polyclonal Antibody

Description
Immunogen sequence: LVSEAIAAGI FNDLGSGSNI DLCVISKNKL DFLRPYTVPN KKGTRLGRYR CEKGTTAVLT EKITPLEIEV LEETVQTM Highest antigen sequence indentity to the following orthologs: Rat - 92%, Mouse - 95%.
Specifications
Specifications
| Antigen | PSMB7 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Psmb7 |
| Gene Accession No. | Q99436 |
| Gene Alias | AU020723; beta-type proteasome 7 subunit; EGK_07890; hypothetical protein LOC511207; macropain chain Z; MC14; Mmc14; Multicatalytic endopeptidase complex chain Z; proteasome (prosome, macropain) subunit, beta type 7; proteasome (prosome, macropain) subunit, beta type, 7; proteasome 20S subunit beta 7; proteasome beta 7 subunit; proteasome catalytic subunit 2; proteasome subunit alpha; proteasome subunit beta 7; proteasome subunit beta type-7; proteasome subunit beta type-7 proprotein; proteasome subunit Z; protein serine kinase c17; Psmb7; RP11-101K10.7; Z |
| Gene Symbols | Psmb7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.