Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSMB8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118206
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118206 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5118206 Supplier Invitrogen™ Supplier No. PA5118206
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PSMB8 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP

The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the proteasome B-type family, also known as the T1B family, that is a 20S core beta subunit. This gene is located in the class II region of the MHC (major histocompatibility complex). Expression of this gene is induced by gamma interferon and this gene product replaces catalytic subunit 3 (proteasome beta 5 subunit) in the immunoproteasome. Proteolytic processing is required to generate a mature subunit. Two alternative transcripts encoding two isoforms have been identified; both isoforms are processed to yield the same mature subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMB8
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Psmb8
Gene Accession No. P28062
Gene Alias ALDD; D6S216; D6S216E; DAAP-57C1.3; JMP; large multifunctional protease 7; Lmp7; Lmp-7; low molecular mass polypeptide 7; low molecular mass protein 7; low molecular weight protein 7; macropain subunit C13; Mc13; MC13 (LMP7); MGC1491; multicatalytic endopeptidase complex subunit C13; NKJO; OTTHUMP00000029420; OTTHUMP00000029421; OTTHUMP00000062981; protease component C13; proteasome (prosome, macropain) subunit, beta type 8 (large multifunctional peptidase 7); proteasome (prosome, macropain) subunit, beta type, 8; proteasome (prosome, macropain) subunit, beta type, 8 (large multifunctional peptidase 7); proteasome catalytic subunit 3i; proteasome component C13; proteasome subunit beta 5i; proteasome subunit beta 8; proteasome subunit beta type 8; proteasome subunit beta type-8; Proteasome subunit beta-5i; proteasome subunit MC13; proteasome subunit Y2; proteasome-related gene 7; proteosome (prosome, macropain) subunit, beta type 8 (large multifunctional peptidase 7); proteosome beta 8 subunit; PSMB5i; Psmb8; Really interesting new gene 10 protein; Ring10; Y2
Gene Symbols Psmb8
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the C-terminus of the Human PSMB8.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5696
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.