Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMB9 Monoclonal Antibody (OTI2B6), TrueMAB™, OriGene
SDP

Catalog No. 502540347
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0347 30 μL
50-167-5101 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0347 Supplier OriGene Supplier No. TA504392S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PSMB9 Monoclonal antibody specifically detects PSMB9 in Canine, Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen PSMB9
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2B6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Psmb9
Gene Accession No. P28065, P28076, P28077
Gene Alias beta1i; large multifunctional peptidase 2; large multifunctional protease 2; LMP2; Lmp-2; LMP-2d; low molecular mass polypeptide 2; low molecular mass protein 2; Macropain chain 7; MGC70470; multicatalytic endopeptidase complex chain 7; proteasome (prosome, macropain) subunit, beta type 9 (large multifunctional peptidase 2); proteasome (prosome, macropain) subunit, beta type, 9; proteasome (prosome, macropain) subunit, beta type, 9 (large multifunctional peptidase 2); proteasome 20S subunit beta 9; proteasome beta 9 subunit; proteasome catalytic subunit 1i; proteasome chain 7; proteasome subunit beta 6i; proteasome subunit beta 9; proteasome subunit beta type 9; proteasome subunit beta type-9; Proteasome subunit beta-1i; proteasome-related gene 2; proteosome (prosome, macropain) subunit, beta type 9; proteosome (prosome, macropain) subunit, beta type 9 (large multifunctional peptidase 2); proteosome beta 9 subunit; PSMB6i; PSMB9; Really interesting new gene 12 protein; Ring12; RING12 protein
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 21-219 of human PSMB9 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16912, 24967, 474867, 5698
Target Species Canine, Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.