Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMB9 Monoclonal Antibody (OTI3H1), TrueMAB™, OriGene
SDP

Catalog No. 502540350
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0350 30 μL
50-167-5097 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0350 Supplier OriGene Supplier No. TA504388S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PSMB9 Monoclonal antibody specifically detects PSMB9 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen PSMB9
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3H1
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SSX5
Gene Accession No. O60225
Gene Alias beta1i; large multifunctional peptidase 2; large multifunctional protease 2; LMP2; Lmp-2; LMP-2d; low molecular mass polypeptide 2; low molecular mass protein 2; Macropain chain 7; MGC70470; multicatalytic endopeptidase complex chain 7; proteasome (prosome, macropain) subunit, beta type 9 (large multifunctional peptidase 2); proteasome (prosome, macropain) subunit, beta type, 9; proteasome (prosome, macropain) subunit, beta type, 9 (large multifunctional peptidase 2); proteasome 20S subunit beta 9; proteasome beta 9 subunit; proteasome catalytic subunit 1i; proteasome chain 7; proteasome subunit beta 6i; proteasome subunit beta 9; proteasome subunit beta type 9; proteasome subunit beta type-9; Proteasome subunit beta-1i; proteasome-related gene 2; proteosome (prosome, macropain) subunit, beta type 9; proteosome (prosome, macropain) subunit, beta type 9 (large multifunctional peptidase 2); proteosome beta 9 subunit; PSMB6i; PSMB9; Really interesting new gene 12 protein; Ring12; RING12 protein
Host Species Mouse
Immunogen Full length human recombinant protein of human SSX5 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6758
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.