Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSMC1 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP214856AF488
Description
PSMC1 Polyclonal antibody specifically detects PSMC1 in Human, Mouse samples. It is validated for Western BlotSpecifications
PSMC1 | |
Polyclonal | |
Western Blot | |
26S protease regulatory subunit 4, 26S proteasome AAA-ATPase subunit RPT2, MGC24583, P26S4, p56, proteasome (prosome, macropain) 26S subunit, ATPase, 1, proteasome 26S ATPase subunit 1, Proteasome 26S subunit ATPase 1, proteasome 26S subunit, ATPase, 1, S4MGC8541 | |
This antibody maps to a region between residue 50 and 100 of human Proteasome (prosome, macropain) Subunit, ATPase, 1 using the numbering given in entry NP_002793.2 (GeneID 5700). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
5700 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction