Learn More
Invitrogen™ PSMC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590640
Description
Immunogen sequence: EFIRNQEQMK PLEEKQEEER SKVDDLRGTP MSVGTLEEII DDNHAIVSTS VGSEHYVSIL SFVDKDLLEP GCSVLLNHKV HAVIGVLMDD TDPLVTVMKV EKAPQETYAD IGGLDNQIQE IKESVELPLT HPEYYEEMGI KPPKGVILYG PPGTGKTLLA KAVANQTSAT FLRVVGSELI QKYLGDGPKL VRELFRVAEE HAPSIVFIDE IDAIGTKRYD SNSGGEREIQ RTMLELLNQL DGFDSRGDVK VIMATNRIET LDPALIRPGR IDRKIEFPLP DEKTKKRIFQ IHTSRMTLAD DVTLDDLIMA KDDLSGADIK AICTEAGLMA LRERRMKVTN EDFKKSKENV LYKKQEGTPE GLYL; Positive Samples: A-549, DU145, Jurkat, HeLa, Mouse brain, Mouse heart, Mouse kidnay, Rat liver; Cellular Location: Cytoplasm, Lipid-anchor, Membrane, Nucleus.
The 26S protease is involved in the ATP-dependent degradation of ubiquitinated proteins. The regulatory (or ATPase) complex confers ATP dependency and substrate specificity to the 26S complex.
Specifications
PSMC1 | |
Polyclonal | |
Unconjugated | |
Psmc1 | |
26S protease regulatory subunit 4; 26S proteasome AAA-ATPase subunit RPT2; 26S proteasome regulatory subunit 4; AI325227; MGC24583; MGC8541; P26s4; p56; peptidase (prosome, macropain) 26S subunit, ATPase 1; protease (prosome, macropain) 26S subunit, ATPase 1; proteasome (prosome, macropain) 26S subunit, ATPase, 1; proteasome 26S ATPase subunit 1; proteasome 26S subunit ATPase 1; proteasome 26S subunit, ATPase 1; proteasome 26S subunit, ATPase, 1; prs4; Psmc1; rpt2; Rpt2/S4; S4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
117263, 19179, 5700 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.77 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
P62191, P62192, P62193 | |
Psmc1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 77-440 of human PSMC1 (NP_0027932). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.