Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSMC1 Polyclonal Antibody

Catalog No. PIPA5121665
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121665 20 μL
PIPA5119362 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121665 Supplier Invitrogen™ Supplier No. PA5121665
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PSMC1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

The 26S protease is involved in the ATP-dependent degradation of ubiquitinated proteins. The regulatory (or ATPase) complex confers ATP dependency and substrate specificity to the 26S complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMC1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene Psmc1
Gene Accession No. P62191, P62192
Gene Alias 26S protease regulatory subunit 4; 26S proteasome AAA-ATPase subunit RPT2; 26S proteasome regulatory subunit 4; AI325227; MGC24583; MGC8541; P26s4; p56; peptidase (prosome, macropain) 26S subunit, ATPase 1; protease (prosome, macropain) 26S subunit, ATPase 1; proteasome (prosome, macropain) 26S subunit, ATPase, 1; proteasome 26S ATPase subunit 1; proteasome 26S subunit ATPase 1; proteasome 26S subunit, ATPase 1; proteasome 26S subunit, ATPase, 1; prs4; Psmc1; rpt2; Rpt2/S4; S4
Gene Symbols Psmc1
Host Species Rabbit
Immunogen Recombinant protein Proteasome 26S Subunit, ATPase 1. The antigen corresponds to amino acid range 1-440 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19179, 5700
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.