Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSMC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA1965
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1965 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1965 Supplier Invitrogen™ Supplier No. PA1965
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

PA1-965 detects proteasome 19S subunit S4 from human cells. PA1-965 has been successfully used in Western blot and immunoprecipitation procedures. By Western blot, this antibody detects a 56 kDa protein representing proteasome 19S subunit S4 in HeLa cell lysate. This antibody detects, to a lesser extent, an ~35 kDa protein which could correspond to subunit degradation product. PA1-965 antigen is yeast proteasome 19S subunit S4.

The 26S protease is involved in the ATP-dependent degradation of ubiquitinated proteins. The regulatory (or ATPase) complex confers ATP dependency and substrate specificity to the 26S complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMC1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene Psmc1
Gene Accession No. P62191
Gene Alias 26S protease regulatory subunit 4; 26S proteasome AAA-ATPase subunit RPT2; 26S proteasome regulatory subunit 4; AI325227; MGC24583; MGC8541; P26s4; p56; peptidase (prosome, macropain) 26S subunit, ATPase 1; protease (prosome, macropain) 26S subunit, ATPase 1; proteasome (prosome, macropain) 26S subunit, ATPase, 1; proteasome 26S ATPase subunit 1; proteasome 26S subunit ATPase 1; proteasome 26S subunit, ATPase 1; proteasome 26S subunit, ATPase, 1; prs4; Psmc1; rpt2; Rpt2/S4; S4
Gene Symbols Psmc1
Host Species Rabbit
Immunogen Yeast proteasome 19S subunit S4.
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5700
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.