Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMC1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987385
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7385 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7385 Supplier Bioss Supplier No. BS6627R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The 26S protease is involved in the ATP-dependent degradation of ubiquitinated proteins. The regulatory (or ATPase) complex confers ATP dependency and substrate specificity to the 26S complex.

Specifications

Antigen PSMC1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Psmc1
Gene Accession No. P62191
Gene Alias 26S protease regulatory subunit 4; 26S proteasome AAA-ATPase subunit RPT2; 26S proteasome regulatory subunit 4; AI325227; MGC24583; MGC8541; P26s4; p56; peptidase (prosome, macropain) 26S subunit, ATPase 1; protease (prosome, macropain) 26S subunit, ATPase 1; proteasome (prosome, macropain) 26S subunit, ATPase, 1; proteasome 26S ATPase subunit 1; Proteasome 26S subunit ATPase 1; proteasome 26S subunit, ATPase 1; proteasome 26S subunit, ATPase, 1; Psmc1; rpt2; Rpt2/S4; S4
Gene Symbols Psmc1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PSMC1/Proteasome 19S S4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5700
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.