Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596024
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596024 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596024 Supplier Thermo Scientific Supplier No. PA596024
Add to Cart
Add to Cart

PSMC2 Polyclonal Antibody

PSMC2 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMC2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.84 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PSMC2
Gene Accession No. P35998, P46471, Q63347
Gene Alias 26S protease regulatory subunit 7; 26S proteasome AAA-ATPase subunit RPT1; 26S proteasome regulatory subunit 7; mammalian suppressor of sgv-1 of yeast; MGC3004; MSS1; Nbla10058; protease 26S subunit 7; proteasome (prosome, macropain) 26S subunit, ATPase 2; proteasome (prosome, macropain) 26S subunit, ATPase, 2; proteasome 26S subunit ATPase 2; proteasome 26S subunit, ATPase 2; proteasome 26S subunit, ATPase, 2; Protein MSS1; PSMC2; putative protein product of Nbla10058; S7; testis secretory sperm-binding protein Li 197a
Gene Symbols PSMC2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human PSMC2 (NP_0027941).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19181, 25581, 5701
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.