Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSMC3 Antibody (OTI9F3), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273696PE
Description
PSMC3 Monoclonal specifically detects PSMC3 in Human, Mouse, Rat, Canine samples. It is validated for Flow Cytometry.Specifications
PSMC3 | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
PSMC3 | |
Full length recombinant protein of human PSMC3 (NP_002795) produced in HEK293T cells. | |
0.1 mL | |
5702.0 | |
Store at 4°C in the dark. |
Flow Cytometry | |
OTI9F3 | |
Flow Cytometry | |
human immunodeficiency virus tat transactivator binding protein-1, MGC8487, proteasome (prosome, macropain) 26S subunit, ATPase, 3,26S proteasome AAA-ATPase subunit RPT5, Proteasome 26S subunit ATPase 3, Proteasome subunit P50, TBP-126S protease regulatory subunit 6A, TBP1Tat-binding protein 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat, Canine | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction