Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSMC3 Monoclonal Antibody (OTI1C10)
GREENER_CHOICE

Catalog No. PIMA525047
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525047 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525047 Supplier Invitrogen™ Supplier No. MA525047
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PSMC3 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

This gene encodes a WASP interacting protein (WIP)-related protein. It has been shown that this protein has a role in the WASP-mediated organization of the actin cytoskeleton and that this protein is a potential link between the activated platelet-derived growth factor receptor and the actin polymerization machinery.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMC3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI1C10
Concentration 0.81 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PSMC3
Gene Accession No. P17980, Q63569
Gene Alias 26S protease regulatory subunit 6A; 26S proteasome AAA-ATPase subunit RPT5; 26S proteasome regulatory subunit 6A; human immunodeficiency virus tat transactivator binding protein-1; MGC8487; proteasome (prosome, macropain) 26S subunit, ATPase 3; proteasome (prosome, macropain) 26S subunit, ATPase, 3; proteasome 26S subunit ATPase 3; proteasome 26S subunit, ATPase 3; proteasome subunit P50; Psmc3; rpt5; Spermatogenic cell/sperm-associated Tat-binding protein homolog SATA; Tat binding protein 1; Tat-binding protein 1; TBP1; TBP-1; testicular secretory protein Li 42
Gene Symbols PSMC3
Host Species Mouse
Immunogen Full length human recombinant protein of PSMC3 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29677, 475980, 5702
Target Species Human, Rat, Canine, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.