Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMC3 Monoclonal Antibody (OTI6E10), TrueMAB™, OriGene
SDP

Catalog No. 502542685
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2685 30 μL
50-167-2064 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2685 Supplier OriGene Supplier No. TA500923S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PSMC3 Monoclonal antibody specifically detects PSMC3 in Canine, Human samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen PSMC3
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI6E10
Concentration 0.79 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PSMC3
Gene Accession No. P17980
Gene Alias 26S protease regulatory subunit 6A; 26S proteasome AAA-ATPase subunit RPT5; 26S proteasome regulatory subunit 6A; human immunodeficiency virus tat transactivator binding protein-1; MGC8487; proteasome (prosome, macropain) 26S subunit, ATPase 3; proteasome (prosome, macropain) 26S subunit, ATPase, 3; proteasome 26S subunit ATPase 3; proteasome 26S subunit, ATPase 3; proteasome subunit P50; Psmc3; rpt5; Spermatogenic cell/sperm-associated Tat-binding protein homolog SATA; Tat binding protein 1; Tat-binding protein 1; TBP1; TBP-1; testicular secretory protein Li 42
Host Species Mouse
Immunogen Full length human recombinant protein of human PSMC3 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 475980, 5702
Target Species Canine, Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.