Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSMC3IP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100560
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100560 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100560 Supplier Invitrogen™ Supplier No. PA5100560
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total HOP2.

PSMC3IP plays an important role in meiotic recombination. It stimulates DMC1-mediated strand exchange required for pairing homologous chromosomes during meiosis. The complex PSMC3IP/MND1 binds DNA, stimulates the recombinase activity of DMC1 as well as DMC1 D-loop formation from double-strand DNA. This complex stabilizes presynaptic RAD51 and DMC1 filaments formed on single strand DNA to capture double-strand DNA. This complex stimulates both synaptic and presynaptic critical steps in RAD51 and DMC1-promoted homologous pairing. It may inhibit HIV-1 viral protein TAT activity and modulate the activity of proteasomes through association with PSMC3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMC3IP
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PSMC3IP
Gene Accession No. Q9P2W1
Gene Alias C79099; DBD-interacting; Gt198; Homologous-pairing protein 2 homolog; homologous-pairing protein 2 homolog-like; Hop2; HUMGT198A; Nuclear receptor coactivator GT198; ODG3; proteasome (prosome, macropain) 26S subunit, ATPase 3, interacting protein; proteasome 26S ATPase subunit 3-interacting protein; PSMC3 interacting protein; PSMC3-interacting protein; PSMC3IP; Tat-binding protein 1-interacting protein; TBP-1-interacting protein; Tbpip
Gene Symbols PSMC3IP
Host Species Rabbit
Immunogen A synthesized peptide derived from human PSMC3IP(Accession Q9P2W1), corresponding to amino acid residues R166-D216.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29893
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.