Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSMC6 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299613AF350
Description
PSMC6 Polyclonal specifically detects PSMC6 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
PSMC6 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
5706 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunofluorescence | |
Alexa Fluor 350 | |
26S protease regulatory subunit 10B, 26S protease regulatory subunit S10B, 26S proteasome AAA-ATPase subunit RPT4, CADP44, conserved ATPase domain protein 44, MGC12520, p42, P44, proteasome (prosome, macropain) 26S subunit, ATPase, 6, Proteasome 26S subunit ATPase 6, Proteasome subunit p42, SUG2 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human PSMC6. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction