Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSMC6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598096
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598096 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598096 Supplier Invitrogen™ Supplier No. PA598096
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PSMC6 Polyclonal Antibody for Western Blot, IHC (P), ELISA

PSMC6 (proteasome (prosome, macropain) 26S subunit, ATPase 6), also known as P44, p42, SUG2, S10B or CADP44, is a regulatory component of the 26S proteasome. More specifically, PSMC6 is one of the six ATPase subunits of the 19S regulator base. It contains a leucine zipper motif and an AAA (ATPase associated with diverse cellular activities) domain and belongs to the AAA ATPase family of chaperone-like ATPases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMC6
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PSMC6
Gene Accession No. P62333, P62334
Gene Alias 2300001E01Rik; 26S protease regulatory subunit 10B; 26S protease regulatory subunit S10B; 26S proteasome AAA-ATPase subunit RPT4; 26S proteasome regulatory subunit 10B; AI451058; CADP44; conserved ATPase domain protein 44; epididymis secretory protein Li 73; HEL-S-73; p42; P44; proteasome (prosome, macropain) 26S subunit, ATPase, 6; proteasome (prosome, macropain) 26S subunit, ATPase, 6-like; proteasome 26S subunit ATPase 6; proteasome 26S subunit, ATPase 6; proteasome subunit p42; PSMC6; SUG2
Gene Symbols PSMC6
Host Species Rabbit
Immunogen Recombinant Human 26S proteasome regulatory subunit 10B protein (1-260AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 289990, 5706, 67089
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.