Learn More
Invitrogen™ PSMC6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144647
Description
Positive control: HepG2 cell lysate, PC-12 cell lysate, rat kidney tissue lysate, 293T cell lysate, SH-SY5Y cell lysate, mouse heart tissue lysate, mouse kidney tissue lysate, SH-SY5Y, rat hippocampus tissue, rat brain tissue, human kidney clear cell carcinoma tissue. Predicted band size: 44 kDa Subcellular Location: Nucleus, Cytoplasm.
PSMC6 (proteasome (prosome, macropain) 26S subunit, ATPase 6), also known as P44, p42, SUG2, S10B or CADP44, is a regulatory component of the 26S proteasome. More specifically, PSMC6 is one of the six ATPase subunits of the 19S regulator base. It contains a leucine zipper motif and an AAA (ATPase associated with diverse cellular activities) domain and belongs to the AAA ATPase family of chaperone-like ATPases.
Specifications
PSMC6 | |
Polyclonal | |
Unconjugated | |
PSMC6 | |
2300001E01Rik; 26S protease regulatory subunit 10B; 26S protease regulatory subunit S10B; 26S proteasome AAA-ATPase subunit RPT4; 26S proteasome regulatory subunit 10B; AI451058; CADP44; conserved ATPase domain protein 44; epididymis secretory protein Li 73; HEL-S-73; p42; P44; proteasome (prosome, macropain) 26S subunit, ATPase, 6; proteasome (prosome, macropain) 26S subunit, ATPase, 6-like; proteasome 26S subunit ATPase 6; proteasome 26S subunit, ATPase 6; proteasome subunit p42; PSMC6; SUG2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
289990, 5706, 67089 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P62333, P62334 | |
PSMC6 | |
Recombinant protein within Human PSMC6 aa 1-200/389. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.