Learn More
PSMC6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 158391AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-0520
Only null left
Add to Cart
Description
Description
PSMC6 (proteasome (prosome, macropain) 26S subunit, ATPase 6), also known as P44, p42, SUG2, S10B or CADP44, is a regulatory component of the 26S proteasome. More specifically, PSMC6 is one of the six ATPase subunits of the 19S regulator base. It contains a leucine zipper motif and an AAA (ATPase associated with diverse cellular activities) domain and belongs to the AAA ATPase family of chaperone-like ATPases.
Specifications
Specifications
PSMC6
Polyclonal
Unconjugated
PSMC6
CADP44, p42, P44, Proteasome subunit p42, PSMC6, Rpt4, SUG2
Rabbit
Antigen Affinity Chromatography
RUO
289990, 5706, 67089
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P62333, P62334
Psmc6
PSMC6 Fusion Protein Ag8589
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.