Learn More
Invitrogen™ PSMD10 Polyclonal Antibody

Description
Antibody detects endogenous levels of total PSMD10.
Specifications
Specifications
| Antigen | PSMD10 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | PSMD10 |
| Gene Accession No. | O75832, Q9Z2X2, Q9Z2X3 |
| Gene Alias | 26S proteasome non-ATPase regulatory subunit 10; 26S proteasome regulatory subunit p28; ankyrin repeat protein; AW554874; dJ889N15.2; Gankyrin; hepatocellular carcinoma-associated protein p28-II; p28; p28(GANK); p28Gank; proteasome (prosome, macropain) 26S subunit, non-ATPase, 10; proteasome 26S subunit, non-ATPase 10; proteasome subunit p28; proteosome (prosome, macropain) 26S subunit, non-ATPase, 10; proteosome subunit p28; PSMD10 |
| Gene Symbols | PSMD10 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.