Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSMD11 Antibody (1F4), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP130252UV
Description
PSMD11 Monoclonal specifically detects PSMD11 in Human, Mouse, C. elegans samples. It is validated for Western Blot, ELISA.Specifications
PSMD11 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PSMD11 | |
Recombinant human PSMD11 (1-422aa) purified from E. coli (NP_002806). | |
0.1 mL | |
Primary | |
This antibody is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human PSMD11 protein. | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, ELISA | |
1F4 | |
Western Blot, ELISA | |
MGC3844,26S proteasome regulatory subunit RPN6, p44.5,26S proteasome regulatory subunit 9, proteasome (prosome, macropain) 26S subunit, non-ATPase, 11,26S proteasome regulatory subunit p44.5, Rpn6,26S proteasome regulatory subunit S9, S9,26S proteasome non-ATPase regulatory subunit 11 | |
Mouse | |
Protein A or G purified | |
RUO | |
5717 | |
Human, Mouse, C. elegans | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction