Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSMD11 Antibody (2C7), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259484F
This item is not returnable.
View return policy
Description
PSMD11 Monoclonal antibody specifically detects PSMD11 in Human samples. It is validated for Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
PSMD11 | |
Monoclonal | |
FITC | |
MGC3844,26S proteasome regulatory subunit RPN6, p44.5,26S proteasome regulatory subunit 9, proteasome (prosome, macropain) 26S subunit, non-ATPase, 11,26S proteasome regulatory subunit p44.5, Rpn6,26S proteasome regulatory subunit S9, S9,26S proteasome non-ATPase regulatory subunit 11 | |
Recombinant human PSMD11 (1-422aa) purified from E. coli | |
0.1 mL | |
Primary | |
Human | |
Purified |
Flow Cytometry, ELISA, Immunofluorescence | |
2C7 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
5717 | |
Store at 4°C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction