Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSMD11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144656
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144656 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144656 Supplier Invitrogen™ Supplier No. PA5144656
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: 293T cell lysate, HepG2 cell lysate, Jurkat cell lysate, NIH/3T3 cell lysate, PC-3 cell lysate, Hela cell lysate, mouse testis tissue lysate, rat testis tissue lysate, mouse brain tissue lysate, rat brain tissue lysate, mouse testis tissue, NIH/3T3. Predicted band size: 47 kDa Subcellular Location: Cytoplasm, Nucleus.

The 26S proteasome (PSMD11) is a multicatalytic proteinase complex with a highly ordered structure composed of 2 complexes, a 20S core and a 19S regulator. The 20S core is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. The 19S regulator is composed of a base, which contains 6 ATPase subunits and 2 non-ATPase subunits, and a lid, which contains up to 10 non-ATPase subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. The protein is a non-ATPase subunit of the 19S regulator.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMD11
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Psmd11
Gene Accession No. F1LMZ8, O00231, Q8BG32
Gene Alias 1700089D09Rik; 1810019E17Rik; 2610024G20Rik; 26S proteasome non-ATPase regulatory subunit 11; 26S proteasome regulatory subunit 9; 26S proteasome regulatory subunit p44.5; 26S proteasome regulatory subunit RPN6; 26S proteasome regulatory subunit S9; 2810055C24Rik; C78232; p44.5; proteasome (prosome, macropain) 26S subunit, non-ATPase, 11; proteasome 26S subunit, non-ATPase 11; Psmd11; Rpn6; S9
Gene Symbols Psmd11
Host Species Rabbit
Immunogen Synthetic peptide within human PSMD11 aa 200-500.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 303353, 5717, 69077
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.