Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSMD13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558497
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558497 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558497 Supplier Invitrogen™ Supplier No. PA558497
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LEKTREKVKS SDEAVILCKT AIGALKLNIG DLQVTKETIE DVEEMLNNLP GVTSVHSRFY DLSSKYYQTI GNHASYYKDA LRFLGCVDIK DLPVSEQQE Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

Acts as a regulatory subunit of the 26S proteasome which is involved in the ATP-dependent degradation of ubiquitinated proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMD13
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PSMD13
Gene Accession No. B0BN93, Q9UNM6, Q9WVJ2
Gene Alias 26S proteasome non-ATPase regulatory subunit 13; 26S proteasome non-ATPase regulatory subunit 13 {ECO:0000250; 26S proteasome regulatory subunit p40.5; 26S proteasome regulatory subunit p40.5 {ECO:0000250; 26S proteasome regulatory subunit RPN9; 26S proteasome regulatory subunit S11; 26S proteasome regulatory subunit S11 {ECO:0000250; 26S proteasome subunit p40.5; EMBL:AAI58733.1, ECO:0000312; EMBL:AAI58733.1}; HSPC027; p40.5; proteasome (prosome, macropain) 26S subunit, non-ATPase, 13; proteasome 26S subunit, non-ATPase 13; PSMD13; psmd13 {ECO:0000312; RGD:1305236}; Rpn9; S11; UniProtKB:Q9UNM6, ECO:0000312; UniProtKB:Q9UNM6}
Gene Symbols PSMD13
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PSMD13. Recombinant protein control fragment (Product #RP-97532).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23997, 365388, 5719
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.