Learn More
Invitrogen™ PSMD13 Polyclonal Antibody

Description
Antibody detects endogenous levels of total PSMD13.
Specifications
Specifications
| Antigen | PSMD13 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | PSMD13 |
| Gene Accession No. | B0BN93, Q9UNM6, Q9WVJ2 |
| Gene Alias | 26S proteasome non-ATPase regulatory subunit 13; 26S proteasome non-ATPase regulatory subunit 13 {ECO:0000250; 26S proteasome regulatory subunit p40.5; 26S proteasome regulatory subunit p40.5 {ECO:0000250; 26S proteasome regulatory subunit RPN9; 26S proteasome regulatory subunit S11; 26S proteasome regulatory subunit S11 {ECO:0000250; 26S proteasome subunit p40.5; EMBL:AAI58733.1, ECO:0000312; EMBL:AAI58733.1}; HSPC027; p40.5; proteasome (prosome, macropain) 26S subunit, non-ATPase, 13; proteasome 26S subunit, non-ATPase 13; PSMD13; psmd13 {ECO:0000312; RGD:1305236}; Rpn9; S11; UniProtKB:Q9UNM6, ECO:0000312; UniProtKB:Q9UNM6} |
| Gene Symbols | PSMD13 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.