Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSMD14 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications
Supplier: Novus Biologicals NBP18095625UL
Description
PSMD14 Polyclonal specifically detects PSMD14 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
PSMD14 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200-1:500 | |
O00487 | |
PSMD14 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:AVDTAEQVYISSLALLKMLKHGRAGVPMEVMGLMLGEFVDDYTVRVIDVFAMPQSGTGVSVEAVDPVFQAKMLDMLKQTGRPEMVVGWYHSHPGFGCWLSGVDINTQQSFEALSERAVAVVVDPIQSVKGKVVIDA | |
25ul | |
Core ESC Like Genes, Stem Cell Markers | |
10213 | |
Rat, Mouse, Human | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
EC 3.1.2.15, EC 3.4.19.-, PAD1, POH126S proteasome non-ATPase regulatory subunit 14,26S proteasome regulatory subunit RPN11, proteasome (prosome, macropain) 26S subunit, non-ATPase, 14, Rpn11, RPN11,26S proteasome-associated PAD1 homolog 1 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of human PSMD14 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction