Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMD2 Monoclonal Antibody (OTI1B5), TrueMAB™, OriGene
SDP

Catalog No. 502541044
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1044 30 μL
50-167-4170 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1044 Supplier OriGene Supplier No. TA503220S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PSMD2 Monoclonal antibody specifically detects PSMD2 in Human samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen PSMD2
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI1B5
Concentration 0.64 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PSMD2
Gene Accession No. Q13200
Gene Alias 26S proteasome non-ATPase regulatory subunit 2; 26S proteasome non-ATPase regulatory subunit 2-like protein; 26S proteasome regulatory subunit RPN1; 26S proteasome regulatory subunit S2; 26S proteasome subunit p97; 55.11 protein; 9430095H01Rik; AA407121; F23F1.8; I79_017809; MGC14274; P97; proteasome (prosome, macropain) 26S subunit, non-ATPase, 2; proteasome 26S subunit, non-ATPase 2; protein 55.11; PSMD2; RPN1; S2; TEG-190; testis expressed gene 190; Tex190; TNFR-associated protein 2; TRAP2; Tumor necrosis factor type 1 receptor-associated protein 2
Host Species Mouse
Immunogen Full length human recombinant protein of human PSMD2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5708
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.