Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSMD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120681
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120681 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120681 Supplier Invitrogen™ Supplier No. PA5120681
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: SW480, OVCAR3, M21, U-937, SH-SY5Y, Mouse testis, Mouse heart, Mouse liver. Immunogen sequence: MEEGGRDKAP VQPQQSPAAA PGGTDEKPSG KERRDAGDKD KEQELSEEDK QLQDELEMLV ERLGEKDTSL YRPALEELRR QIRSSTTSMT SVPKPLKFLR PHYGKLKEIY ENMAPGENKR FAADIISVLA MTMSGERECL KYRLVGSQEE LASWGHEYVR HLAGEVAKEW QELDDAEKVQ REPLLTLVKE IVPYNMAHNA EHEACDLLME IEQVDMLEKD IDENAYAKVC LYLTSCVNYV PEPENSALLR CALGVFRKFS.

The 26S proteasome is a multicatalytic proteinase complex with a highly ordered structure composed of 2 complexes, a 20S core and a 19S regulator. The 20S core is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. The 19S regulator is composed of a base, which contains 6 ATPase subunits and 2 non-ATPase subunits, and a lid, which contains up to 10 non-ATPase subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes one of the non-ATPase subunits of the 19S regulator lid. In addition to participation in proteasome function, this subunit may also participate in the TNF signalling pathway since it interacts with the tumor necrosis factor type 1 receptor. A pseudogene has been identified on chromosome 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMD2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.79 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PSMD2
Gene Accession No. Q13200, Q8VDM4
Gene Alias 26S proteasome non-ATPase regulatory subunit 2; 26S proteasome non-ATPase regulatory subunit 2-like protein; 26S proteasome regulatory subunit RPN1; 26S proteasome regulatory subunit S2; 26S proteasome subunit p97; 55.11 protein; 9430095H01Rik; AA407121; F23F1.8; I79_017809; MGC14274; P97; proteasome (prosome, macropain) 26S subunit, non-ATPase, 2; proteasome 26S subunit, non-ATPase 2; protein 55.11; PSMD2; RPN1; S2; TEG-190; testis expressed gene 190; Tex190; TNFR-associated protein 2; TRAP2; Tumor necrosis factor type 1 receptor-associated protein 2
Gene Symbols PSMD2
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21762, 5708
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.