Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMD3 Monoclonal Antibody (OTI2A2), TrueMAB™, OriGene
SDP

Catalog No. 502540377
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0377 30 μL
50-167-5055 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0377 Supplier OriGene Supplier No. TA504337S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PSMD3 Monoclonal antibody specifically detects PSMD3 in Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen PSMD3
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI2A2
Concentration 0.95 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene FXN
Gene Accession No. Q16595
Gene Alias 26S proteasome non-ATPase regulatory subunit 3; 26S proteasome regulatory subunit RPN3; 26S proteasome regulatory subunit S3; AI255837; AntP91a; D11Bwg1349e; EGK_08646; P58; P91a; proteasome (prosome, macropain) 26S subunit, non-ATPase, 3; proteasome 26S non-ATPase subunit 3; proteasome 26S subunit, non-ATPase 3; proteasome subunit p58; Psd3; Psmd3; RPN3; S3; tissue specific transplantation antigen 2; tissue specific transplantation antigen P91A; transplantation antigen P91A; TSTA2; Tstap91a; Tum-P91A antigen
Host Species Mouse
Immunogen Full length human recombinant protein of human FXN produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2395
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.