Learn More
Invitrogen™ PSMD4 Recombinant Rabbit Monoclonal Antibody (5G1X6)

Description
Immunogen sequence: TTGTEDSDDA LLKMTISQQE FGRTGLPDLS SMTEEEQIAY AMQMSLQGAE FGQAESADID ASSAMDTSEP AKEEDDYDVM QDPEFLQSVL ENLPGVDPNN E.
Specifications
Specifications
| Antigen | PSMD4 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 5G1X6 |
| Concentration | 0.63 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Psmd4 |
| Gene Accession No. | O35226, P55036 |
| Gene Alias | 26S proteasome non-ATPase regulatory subunit 4; 26S proteasome regulatory subunit RPN10; 26S proteasome regulatory subunit S5A; AF; Af1; AF-1; angiocidin; anti secretory factor 1; antisecretory factor; antisecretory factor 1; ASF; Mcb1; multiubiquitin chain-binding protein; multiubiquitin-chain-binding protein; proteasome (prosome, macropain) 26S subunit, non-ATPase, 4; proteasome 26S subunit, non-ATPase 4; Psmd4; pUB-R5; RP11-126K1.1; Rpn10; RPN10 homolog; S5A; S5a/antisecretory factor protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.