Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSMD6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP18154525UL
Description
PSMD6 Polyclonal specifically detects PSMD6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
PSMD6 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:10-1:20 | |
26S proteasome non-ATPase regulatory subunit 6, Breast cancer-associated protein SGA-113M, KIAA010726S proteasome regulatory subunit S10, p42A, p44S10, PFAAP4, Phosphonoformate immuno-associated protein 4, proteasome (prosome, macropain) 26S subunit, non-ATPase, 6,26S proteasome regulatory subunit RPN7, Proteasome regulatory particle subunit p44S10, Rpn7, S10, SGA-113M | |
Rabbit | |
Affinity Purified | |
RUO | |
9861 | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
PSMD6 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:NMAPYYEALCKSLDWQIDVDLLNKMKKANEDELKRLDEELEDAEKNLGESEIRDAMMAKAEYLCRIGDKEGALTAFRKTYDKTVALG | |
25 μL | |
Primary | |
Specificity of human PSMD6 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction