Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSMD6 Polyclonal Antibody
PSMD6 Polyclonal Antibody
Supplier: Thermo Scientific PA530333
Description
PSMD6 Polyclonal Antibody for Western Blot, IHC (P)

Specifications
PSMD6 | |
Polyclonal | |
Unconjugated | |
PSMD6 | |
2400006A19Rik; 26S proteasome non-ATPase regulatory subunit 6; 26S proteasome non-ATPase regulatory subunit 6-like protein; 26S proteasome non-ATPase regulatory subunit-like protein; 26S proteasome regulatory subunit RPN7; 26S proteasome regulatory subunit S10; breast cancer-associated protein SGA-113M; KIAA0107; p42A; P44s10; PFAAP4; Phosphonoformate immuno-associated protein 4; proteasome (prosome, macropain) 26S subunit, non-ATPase, 6; proteasome 26S subunit, non-ATPase 6; proteasome regulatory particle subunit p44S10; proteasome, 26S, non-ATPase regulatory subunit 6; PSMD6; Rpn7; S10; SGA-113M; vov; zgc:56471 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
289924, 66413, 9861 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.87 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q15008, Q99JI4 | |
PSMD6 | |
Recombinant fragment corresponding to a region within amino acids 23 and 307 of Human PSMD6. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction