Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMD6 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989019
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9019 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9019 Supplier Bioss Supplier No. BS9367R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of the protease subunit S10 family. The encoded protein is a subunit of the 26S proteasome which colocalizes with DNA damage foci and is involved in the ATP-dependent degradation of ubiquinated proteins. Alternative splicing results in multiple transcript variants.

Specifications

Antigen PSMD6
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PSMD6
Gene Accession No. Q15008
Gene Alias 2400006A19Rik; 26S proteasome non-ATPase regulatory subunit 6; 26S proteasome regulatory subunit RPN7; 26S proteasome regulatory subunit S10; BOS_20895; Breast cancer-associated protein SGA-113M; KIAA0107; p42A; p44S10; PFAAP4; Phosphonoformate immuno-associated protein 4; proteasome (prosome, macropain) 26S subunit, non-ATPase, 6; proteasome 26S subunit, non-ATPase 6; Proteasome regulatory particle subunit p44S10; proteasome, 26S, non-ATPase regulatory subunit 6; Psmd6; Rpn7; S10; SGA-113M; vov; zgc:56471
Gene Symbols PSMD6
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PSMD6/Proteasome regulatory particle subunit p44S10.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9861
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.