Learn More
PSMD7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA527941
Description
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (98%), Rat (97%), Zebrafish (97%), Xenopus laevis (96%), Rhesus Monkey (99%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Proteolytic degradation is critical to the maintenance of appropriate levels of short-lived and regulatory proteins as important and diverse as those involved in cellular metabolism, heat shock and stress response, antigen presentation, modulation of cell surface receptors and ion channels, cell cycle regulation, transcription, and signaling factors. The ubiquitin-proteasome pathway deconstructs most proteins in the eukaryotic cell cytosol and nucleus. Others are degraded via the vacuolar pathway which includes endosomes, lysosomes, and the endoplasmic reticulum. The 26S proteasome is an ATP-dependent, multisubunit (approximately 31), barrel-shaped molecular machine with an apparent molecular weight of approximately 2.5 MDa. It consists of a 20S proteolytic core complex which is crowned at one or both ends by 19S regulatory subunit complexes. The 19S regulatory subunits recognize ubiquitinated proteins and play an essential role in unfolding and translocating targets into the lumen of the 20S subunit. An enzymatic cascade is responsible for the attachment of multiple ubiquitin molecules to lysine residues of proteins targeted for degradation. Several genetic diseases are associated with defects in the ubiquitin-proteasome pathway. Some examples of affected proteins include those linked to cystic fibrosis, Angelman's syndrome, and Liddle syndrome.
Specifications
| PSMD7 | |
| Polyclonal | |
| Unconjugated | |
| Psmd7 | |
| 26S proteasome non-ATPase regulatory subunit 7; 26S proteasome non-ATPase regulatory subunit 7-like protein; 26S proteasome regulatory subunit rpn8; 26S proteasome regulatory subunit S12; AW107203; Moloney leukemia virus-34 proviral integration; Mov34; Mov-34; Mov34 homolog; Mov34 protein; Mov34 protein homolog; MOV34L; P40; proteasome (prosome, macropain) 26S subunit, non-ATPase, 7; proteasome (prosome, macropain) 26S subunit, non-ATPase, 7 (Mov34 homolog); proteasome 26S non-ATPase subunit 7; proteasome 26S subunit, non-ATPase 7; proteasome subunit p40; Psmd7; Rpn8; S12; wu:fi03c01; zgc:55284 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5713 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Western Blot | |
| 0.78 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| P51665 | |
| Psmd7 | |
| Recombinant fragment corresponding to a region within amino acids 47 and 324 of Human PSMD7 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.